Learn More
Medchemexpress LLC Madecassic acid | 18449-41-7 | 504.70 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN0569100MG
Encompass_Preferred
Madecassic acid is isolated from *Centella asiatica* (Umbelliferae) and possesses anti-inflammatory properties. It works by inhibiting iNOS, COX-2, TNF-alpha, IL-1beta, and IL-6 through the downregulation of NF-κB activation in RAW 264.7 macrophage cells.
  • Isolated from *Centella asiatica*.
  • Possesses anti-inflammatory properties.
  • Inhibits iNOS and COX-2 protein expression.
  • Reduces IL-17 levels and γδT17 cells in colitis mice.
  • Attenuates colitis in mice.
Catalog No. 50-003-88848