Learn More
Medchemexpress LLC Madecassic acid | 18449-41-7 | 99.8% | 504.70 | 10 MG
SDP
Supplier:  Medchemexpress LLC 10MGHYN0569
Encompass_Preferred
Madecassic acid is a compound isolated from Centella asiatica (Umbelliferae). It demonstrates anti-inflammatory properties by inhibiting various inflammatory mediators through the downregulation of NF-κB activation in RAW 264.7 macrophage cells.
  • Isolated from Centella asiatica
  • Exhibits anti-inflammatory properties
  • Inhibits iNOS and COX-2 protein expression
  • Downregulates NF-κB activation
  • Reduces IL-17 levels and γδT17 cells in colitis models
  • For research use only
Catalog No. 50-003-65631