Learn More
Medchemexpress LLC HY-100236 5mg Medchemexpress, Madrasin CAS:374913-63-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1002365MG
Encompass_Preferred
Medchemexpress, HY-100236 5mg Madrasin CAS:374913-63-0 Madrasin (DDD00107587) is a splicing inhibitor that prevents formation of both splicing intermediates and products in vitro and interferes with one or more early steps in the pathway of spliceosome assembly. Madrasin also can inhibit pre-mRNA splicing in vitro and modify splicing of endogenous pre-mRNA in cells [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1770