Learn More
Aviva Systems Biology Corp MAFG Antibody - N-terminal region (ARP38143_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38143_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAFG. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MAFG. Alternate names: MGC13090, MGC20149, hMAFApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-56534