Learn More
Seracare Life Sciences Inc MAG BEADS ANTI-E.COLI O145 1ML
Supplier:  Seracare Life Sciences Inc 53500042
Encompass_Preferred
Anti-E.coli O145 Magnetic Beads are super-paramagnetic polystyrene beads coated with KPL's Anti-E.coli O145 Antibody. They are intended for use in the isolation and separation of E.coli O145 from a variety of samples.
Catalog No. 50-607-727