Learn More
V&P Scientific Inc MAGNETIC RACK, 1 Flip Top Microcentrifuge Tube, 1.5mL Volume
Supplier:  V&P Scientific Inc VP 772F5
Encompass
MAGNETIC BEAD SEPARATION RACK, 1 Flip Top Microcentrifuge Tube, 1.5mL Volume, 1 Sided Bead Pellet Location, High and Low Position, 52MGO, NdFeB Magnet, 11mm Hole Diameter, UHMW
Catalog No. NC0457049