Learn More
Cole-Parmer Lab Companion AAA330115B Round top Magnetic Stirrer with silicon plate cover, 2L, 100-240V
Supplier:  Cole-Parmer 8657507
Encompass
Stirrers come with one black and one white silicone plate cover so you can choose the best contrast for your application. The body is constructed of lightweight polypropylene for easy handling, and the rubber feet stabilize the unit even when in the highest speed setting. Brushless motor provides durability and quiet operation.
Catalog No. NC0485035