Learn More
Bioworld Magentaâ„¢ GA-7-3 Plant Culture Box w/ Lid, 100 Unit(s)
Supplier:  Bioworld 309300102
Encompass
Made of polycarbonate, non-sterile vessel with polypropylene lid. Magenta boxes are vessels used in the plant culture process. The boxes are autoclavable and resuable. They are sturdy and leak free. The clear surface allows one to view samples without disrupting growth. Dimensions: 3 x 3 x 3" Sold as 100/case. Please note:This product's shipping cost is calculated based on the dimensions of the largest package size. All shipping charges will be adjusted internally upon receipt of the order to accurately reflect the standard carrier costs associated with the package sizes ordered.
Catalog No. NC0776391