Learn More
Alkali Scientific Magenta Gal [5 Bromo 6 chloro 3 indolyl β D galactopyranoside], 1 G
SDP
Supplier:  Alkali Scientific AGE002CP
Encompass
Magenta Gal (5 Bromo 6 chloro 3 indolyl β D galactopyranoside) is a chromogenic substrate used in laboratory research for the detection of β-galactosidase activity. This 1 g reagent is useful in assays that require the colorimetric detection of enzyme activity. When β-galactosidase breaks down the substrate, a color change occurs, making it easy to observe and measure enzymatic reactions. It is commonly used in microbiology, molecular biology, and diagnostic research for studies on gene expression, enzyme activity, and cellular metabolism. Magenta Gal is especially valuable for screening experiments where β-galactosidase serves as a reporter enzyme.
Catalog No. 50-006-60155