Learn More
Research Products International Corp MAGic Clamp Adjustable, for 125ml, 250ml, 500ml Flasks
SDP
Supplier:  Research Products International Corp 260128
Encompass
MAGic Clamp Adjustable is designed for use with Incu-Shaker Mini and Incu-Shaker 10L model Shaking Incubators. Holds 125, 250, and 500ml flasks. This unique, magnetic attachment allows easy interchanging between flask clamps of different sizes.
Magnetic flask clamp secures Erlenmeyer flask to a shaker platform for mixing and aerating procedures.
Two year warranty.
Catalog No. 50-222-9813