Learn More
Supply Solutions Magic Rack Universal Platform
Supplier: Supply Solutions BENCHMARK BT3000MR
The powerful Orbi-Shaker is capable of mixing a 14lb load capacity up to speeds as high as 300 RPM. The standard platform (included) features a non-slip, rubber coated surface, ideal for tissue culture flasks, petri dishes and staining trays. The convenient MAGic ClampT platform (PATENT PENDING) is also available for use with Erlenmeyer flasks and test tube racks. The unique, magnetic attachment method is the easiest way to instantly change between flask clamps of different sizes. Dimensions (W x D x H) of BT3000 : 33 x 30 CM (13 x 12 IN) (Included) Dimensions (W x D x H) of BT3000-MR : 35 x 30 CM (14 x 12 IN) (MAGic Clamp) Operating Temperature Range : +4 to +65 C Electrical Requirements : 120V or 230V, 50-60Hz 600W