Learn More
LIFE TECH (BID ECHO ONLY) MagicMark™ XP Western Protein Standard, 250 μL
Supplier: LIFE TECH (BID ECHO ONLY) LC5602
- MagicMark XP Western Protein Standard consists of nine recombinant proteins (20–220 kDa), each of which contains an IgG binding site
- The IgG binding site binds the primary or secondary antibody used for detection of the target protein, allowing direct visualization of the standard on the western blot
- The MagicMark XP Standard is compatible with most western kits and substrates (chemiluminescent, chromogenic, and fluorescent)
- The protein standard is supplied in a ready-to-use format for direct loading onto gels; no need to heat, reduce, or add sample buffer prior to use
- Store at -20°C
Applications
- Western blotting: detection of the nine unstained bands via the detection method used for the target protein. /li>
- Protein size estimation: protein sizing on SDS-polyacrylamide gels and on western blot using western blot detection method
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.