Learn More
Cayman Chemical MAGLhumnrecombInt 1ea
SDP
Supplier:  Cayman Chemical 7051941 EA
Encompass_Preferred
MAGL (human, recombinant) has been tested and formulated to work exclusively with Cayman’s Monoacylglycerol Lipase Inhibitor Screening Assay Kit (Item No. 705192).
Catalog No. 50-289-4590