Learn More
Alkali Scientific Magnesium Chloride 2M Solution, Packaged 5 x 10 Milliliters, 50 Milliliters
SDP
Supplier:  Alkali Scientific ORI001CP
Encompass_Preferred
Magnesium Chloride 2M Solution is a high-concentration chemical reagent commonly used in laboratory applications. This 50mL bottle contains a 2M solution of magnesium chloride, which is often employed in experiments requiring magnesium ions for biochemical reactions. It is widely used in DNA/RNA precipitation, enzyme activation, and protein binding studies. Magnesium chloride plays an important role in stabilizing structures like RNA and DNA during experimental procedures. It is also a key component in various buffer solutions used in cell biology, biochemistry, and microbiology for maintaining the stability of proteins and enzymes. This solution is ideal for preparing working solutions and dilutions in various lab procedures.
Catalog No. 50-006-60997