Learn More
Crescent Chemical Co Inc MAGNESIUM CHLORIDE.6H2O
SDP
Supplier:  Crescent Chemical Co Inc SE39771.01
Encompass_Preferred
Magnesium chloride.6H2O molecular biology grade DNase/RNase not detected. Size - 500G Storage Conditions - +15 °C TO +30 °C Catalog Number - 39771.01 CAS 7791-18-6
Catalog No. 50-248-456