Learn More
New England Biolabs, Inc. NEBNext Magnesium RNA Fragmentation Module – 200 reactions
SDP
Supplier:  New England Biolabs, Inc. E6150S/DEL
Encompass
Fragmentation of RNA can be performed by enzymes, heat alone, or exposure to divalent cations at elevated temperature. This kit provides highly efficient and tunable RNA fragmentation by incubation with Magnesium ions at 94°C.
Catalog No. NC0477295