Learn More
Medchemexpress LLC Magnesium glycinate | 14783-68-7 | 98.0% | 5 G
SDP
Supplier:  Medchemexpress LLC HY1293285G
Encompass_Preferred
Magnesium glycinate is the magnesium salt of glycine and functions as a nutrient supplement. It possesses satisfactory physico-chemical properties and bioactivities. Metal glycinate chelates are formed through chemical reactions between glycine and metal compounds. Magnesium, an essential mineral, plays a critical role in energy metabolism and helps maintain normal muscle function. It is considered an ideal nutrient supplement for the prophylaxis and treatment of metal deficiency.
  • Nutrient supplement
  • Satisfactory physico-chemical properties and bioactivities
  • Formed through chemical reactions between glycine and metal compounds
  • Magnesium plays a critical role in energy metabolism
  • Assists in maintaining normal muscle function
  • Ideal for prophylaxis and treatment of metal deficiency
Catalog No. 50-003-39709