Learn More
Medchemexpress LLC Magnesium Lithospermate B | 122021-74-3 | 99.49% | 740.90 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY12641510MG
Encompass_Preferred
Magnesium Lithospermate B is a derivative of caffeic acid tetramer and an inhibitor of Na+/K+ ATPase, which can be extracted from *Salviae miltiorrhizae*. It is widely used for the research of cardiovascular diseases and can protect against glucose-induced intracellular oxidative damage. It also suppresses neuroinflammation and attenuates neurodegeneration.
  • Decreases LDH activity and increases SOD activity in cardiomyocytes
  • Reduces intracellular ROS and MDA levels, and significantly suppresses cardiomyocytes apoptosis
  • Enhances proliferation of neural stem cells (NSCs) in a dose-dependent manner
  • Promotes the differentiation in vitro of NSCs towards neurons
  • Reduces the renal damage of oxidative stress through reduction of reactive oxygen species in old rats
  • Promotes neurogenesis and improves memory in PD models
Catalog No. 50-004-38192