Learn More
Medchemexpress LLC Magnesium Lithospermate B | 122021-74-3 | 99.49% | 740.90 | 100 MG
Supplier: Medchemexpress LLC HY126415100MG
Magnesium Lithospermate B is a derivative of caffeic acid tetramer and an inhibitor of Na+/K+ ATPase, which can be extracted from *Salviae miltiorrhizae*. It is widely used for the research of cardiovascular diseases and can protect against glucose-induced intracellular oxidative damage. It also suppresses neuroinflammation and attenuates neurodegeneration.
- Decreases LDH activity, increases SOD activity, reduces intracellular ROS and MDA levels, and significantly suppresses cardiomyocytes apoptosis at concentrations of 20-60 μg/ml (24 h).
- Enhances proliferation of neural stem cells (NSCs) in a dose-dependent manner (1-100 μg/ml).
- Promotes the differentiation in vitro of NSCs towards neurons (10 μg/ml).
- Reduces renal damage from oxidative stress by decreasing reactive oxygen species in old rats (2-8 mg/kg; p.o. once daily for 16 d).
- Promotes neurogenesis and improves memory in PD models (0.5 μg/g; s.c. for 6 weeks).