Learn More
Medchemexpress LLC Magnesium silicate (Activated magnesium silicate) | 1343-88-0 | 500 G
SDP
Supplier:  Medchemexpress LLC HYB2205500G
Encompass_Preferred
Magnesium silicate (Activated magnesium silicate) is an orally active compound composed of magnesium oxide (MgO) and silicon dioxide (SiO2). It is used in the preparation of food additives, antacids, and anti-ulcer agents, and also functions as a piezoelectric biomaterial, deodorant, decolorizing agent, and antifungal agent.
  • Releases magnesium and silicon ions
  • Promotes differentiation of mesenchymal stem cells into osteoblasts
  • Improves osteogenic properties when combined with various polymers
  • Can be neutralized in the stomach to form magnesium chloride and silicon dioxide
  • Acts as a glidant and anticaking agent
  • Functions as an antiacid and antiulcer agent
  • Exhibits antifungal properties
  • Can be used as a piezoelectric biomaterial
Catalog No. 50-004-12346