Learn More
Strem, An Ascensus Company CAS# 60871-83-2. 25g. Magnesium trifluoromethanesulfonate, min. 98% (Magnesium triflate). MFCD00042601
SDP
Supplier:  Strem, An Ascensus Company 12400025G
Encompass
CAS# 60871-83-2. 25g. Magnesium trifluoromethanesulfonate, min. 98% (Magnesium triflate). MFCD00042601. Molecular Weight: 322.45. Molecular Formula: Mg(SO3CF3)2. Color/form: white pwdr. Strem# 12-4000. http://www.strem.com/catalog/v/12-4000/
Catalog No. 50-901-09511