Learn More
V&P Scientific Inc STIR ELEMENT, CYLINDER, NdFeB Magnet, 42MGO, PEEK Encapsulated, Used in Bottles, Tubes/Vials, 10/pack
Supplier:  V&P Scientific Inc VP 772BKN421123
Encompass
STIR ELEMENT, CYLINDER, NdFeB Magnet, 42MGO, PEEK Encapsulated, 11.5mm Diameter, 23.1mm Length, 150°C Max Operating Temp for Magnets
Catalog No. NC0671035