Learn More
Research Products International Corp MAG-NET Magnetic Spin Bar Remover/Restrainer
SDP
Supplier:  Research Products International Corp 249010
Encompass
MAG-NET allows removal of spin bar from any beaker without touching the solution. Simply place MAG-NET against side of beaker and slide upwards. Conveniently hold spin bar in place while pouring solution from beaker. Safer and more convenient than immersion style retrievers. Heavy duty magnet is encased in polypropylene for easy cleaning and autoclaving.
Catalog No. 50-550-325