Learn More
World Precision Instrument Magnetic Stand - Compact, 5 to 6.5 mm
SDP
Supplier:  World Precision Instrument MB2
Encompass
The base of each stand exerts a powerful magnetic force that holds it solidly on ferrous metal surfaces - even vertically or upside down
Mechanical clamping type tightens three joints simultaneously just by one-touch operation. Arm is freely adjustable without distortion. Equipped with fine adjuster and medium size magnet for stabilizing the base. Suitable for performing precision operation.
Catalog No. 50-822-975