Learn More
World Precision Instrument M10 Magnetic Stand
SDP
Supplier:  World Precision Instrument M10
Encompass
The base of each stand exerts a powerful magnetic force that holds it solidly on ferrous metal surfaces - even vertically or upside down
This magnetic base is similar to M1 but with a 12mm diameter sub pole that fits the 12mm clamp supplied with M3301, DC3001, MD4 and MMJ manipulators. These are an ideal manipulator stands.
Catalog No. 50-822-962