Learn More
Chemglass Life Sciences Chemglass Stirring Bar, Plain PTFE Magnetic, 20 x 6 mm (For magnetic stir plates)
SDP
Supplier:  Chemglass Life Sciences CG2001P03
Encompass_Preferred
PTFE plain magnetic stirring bar, 20 x 6 mm, designed for use with magnetic stir plates to provide consistent mixing. The plain shape increases turbulence at low speeds compared with smooth cylindrical bars, and the PTFE construction offers broad chemical resistance suitable for common laboratory applications.
  • PTFE construction for chemical resistance.
  • Plain shape increases turbulence at low stirring speeds.
  • 20 x 6 mm size for small to medium vessels.
  • Magnetized for use with magnetic stir plates.
  • Sold individually (each).
Catalog No. NC2609505