Learn More
V&P Scientific Inc STIR ELEMENT, CYLINDER, NdFeB Magnet, 52MGO, PEEK Encapsulated, Used in Bottles, 10/pack
Supplier:  V&P Scientific Inc VP 772BKN521148
Encompass
STIR ELEMENT, CYLINDER, NdFeB Magnet, 52MGO, PEEK Encapsulated, 11.5mm Diameter, 48.2mm Length, 80°C Max Operating Temp for Magnets
Catalog No. NC1916684