Learn More
Sigma Aldrich Fine Chemicals Biosciences Magnetic Stir Bar round wh
Supplier:  Sigma Aldrich Fine Chemicals Biosciences HS12054910EA
Encompass
The ALNICO magnetic stir bar withstands temperatures up to 250C making them suitable for use with both unheated and heated stirrers. Low friction octagon shape with a center pivot ring helps create a strong mixing vortex even with more viscous SOLUTIONons
Catalog No. 50-290-1722