Learn More
V&P Scientific Inc STIR ELEMENT, DISC, NdFeB Magnet, 42MGO, PVDF Encapsulated, Used in Microplates, Syringes, Tubes/Vials, 10/pack
Supplier:  V&P Scientific Inc VP 772DPN4252
Encompass
STIR ELEMENT, DISC, NdFeB Magnet, 42MGO, PVDF Encapsulated, 5.0mm Diameter, 1.7mm Height, 150°C Max Operating Temp for Magnets
Catalog No. NC0947407