Learn More
Sigma Aldrich Fine Chemicals Biosciences Magnetic stirrer battery p1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences BAH3701700001EA
Encompass
Battery powered magnetic stirrer offers portable lightweight stirring power.Ideal for field use inside incubators and anywhere electricity is not convenient or available Operates on 1 or 2 D batteries 2 batteries will provide approximately 16 days of continuous operation Mix up to 1L Speed control knob with separate on/off switch operate up to 900RPM Durable chemical-resistant polypropylene
Catalog No. 50-290-5515