Learn More
Bioworld Concanavalin A (Jackbean) Lectin (Con A) - MagneZoomâ„¢ Kit
Supplier:  Bioworld 201630201
Encompass
Technical Specifications:Ligand Source:Canavalia ensiformisMatrix: MagneZOOM™ (Paramagnetic Silica Coated Beads)Particle size range: 2-5 µmMatrix attachment: AmideBinding Capacity: 1-2 mg/mLInhibiting/Eluting Sugar:a-Methylglucoside +a-MethylmannosideCarbohydrate for Elution: 0.2Ma-Methylglucoside +a-MethylmannosideBlood Group Specificity: N/AForm: 25 mg/mL , PBS (1x) pH 7.4 w/ 0.02% sodium azidepH stability: 4 – 9Storage: 2 – 8 °C, DO NOT FREEZEApplication note: Calcium and Manganese are required for binding. Kit contents:• MagneZoom™-ConA, 25mg/ml suspension: 2 mL• Binding/Wash Buffer • Elution Buffer• Storage BufferThis Kit Requires MagneZoom paramagnetic bead station (20160000-1) for magnetic bead separation.
Catalog No. 50-197-623