Learn More
Bioworld MagneZoom™ Goat Anti-Mouse IgG (FC) Antibody Kit (Polyclonal IgG), 2 mL
Supplier:  Bioworld 201630171
Encompass
MagneZoom ™ Goat Anti-Mouse IgG (FC) Antibody Kit (Polyclonal IgG)Cat#20163017DetailsHost:GoatIsotype:IgGClass:PolyclonalImmunogen:Mouse IgG (FC)Form InformationForm:Liquid SuspensionConcentration:25 mg MagneZoom™ per mL of bufferPurification:Protein A ChromatrographyStorage Buffer:PBS with 0.1% BSAPreservative:Upon Request (0.02% Sodium Azide)Storage:4°CKit contents:• MagneZoom Goat Anti-mouse IgG(Fc), 25mg/ml suspension magnetic beads: 2ml• Binding/Wash Buffer •Elution Buffer• Neutralization Buffer
Catalog No. 50-197-621