Learn More
Bioworld MagneZoom™ G (Protein G), 5 mL
Supplier:  Bioworld 201620053
Encompass
Recombinant protein G covalently linked to MagneZoom (paramagnetic beads). Protein G binds Rc region of mammalian immunoglobulin. Ideal for purification and fractionation of IgG subclasses that poorly bind protein A. Suitable for high throughput screening.Technical SpecificationsParticle size: 2-5µmLigand: rProtein GBinding capacity: ~200µg IgG/ml beadspH stability: 4-9Supplied as 5mg/ml suspensionRequires MagneZoom paramagnetic bead station (20160000-1) for separating beads.Use: bioWorld's products are forRESEARCH use only.
Catalog No. 50-197-585