Learn More
Bioworld MagneZoom™ Oligo (dT)25, 2 mL
Supplier:  Bioworld 201600031
Encompass
For isolation and purification of pure and intact mRNA from crude extract of animals and plant tissues. It is supplied as a suspension of super paramagnetic beads covalently coupled to Oligo (dT)25.Technical specification:Matrix: Silica coated Magnetic beadsLigand: Oligo (dT)25Binding Capacity: 2µg of mRNA/mg beads (10µg mRNA/1ml suspension)Particale size: 2-5µmSupplied as: 5mg/ml suspensionin 0.25M Tris, 0.02M EDTA, 0.1% Tween 20 and 0.02% Sodium AzideRequires MagneZoom™ paramagnetic bead station (20160000-1).
Catalog No. 50-197-560