Learn More
Dazor Mfg Corporation LED Hi-Lighting Desk Base Magnifier 28" Arm Reach, w/ 3 Diopter 5" Lens, 1.75X Magnification, Color: Black
Supplier:  Dazor Mfg Corporation LMG400BK
Encompass_Preferred
Dazor's LED Hi-Lighting Magnifier is the ideal magnifier for viewing details on uneven surfaces. Exact positioning of the light source behind the lens casts the light at an angle creating shadows to emphasis contrasts on irregular surfaces. The Daylight illumination color and quality promotes accurate viewing when observing subtle differences in surfaces. Features: Exact positioning of the 72 fully diffused LEDs behind the lens casts the light at an angle creating a highlighting effect to emphasis contrasts on irregular surfaces. 6500K Daylight with 92+ CRI for superior viewing and color accuracy. Clear crown-optical glass lens for clear and distortion-free viewing.Features Dazor's patented 28" contemporary-style floating arm for smooth and easy positioning of the lamp to any position. Variable intensity dimming knob. Sturdy weighted desk base. Durable powder-coat finish. 5 year limited warranty. Made in the USA.
Catalog No. NC1580532