Learn More
TARGETMOL CHEMICALS INC Magnolol 100MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T3000100MG
Encompass_Preferred
Also available in 1 g, 1 mL, 10 mg, 25 mg, 50 mg, 500 mg and bulk. Please contact Fisher for quotes. Magnolol (5,5'-Diallyl-2,2'-biphenyldiol) is a dual agonist of RXRalpha( EC50=10.4 uM) and PPARgamma(EC50=17.7 uM). It blocks TNF-alpha-induced NF-KB activation. Purity 100%
Catalog No. 50-287-9305