Learn More
Apexbio Technology LLC Magnolol 528-43-8 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC N13595.1
Encompass_Preferred
Magnolol is a natural biphenolic compound structurally related to honokiol exhibiting inhibitory activity on the production of nitric oxide (NO) and TNF- It functions by suppressing inducible nitric oxide synthase (iNOS) expression in macrophages activated by lipopolysaccharide (LPS) Magnolol demonstrates antifungal and antibacterial activities and is also reported to possess anxiolytic and anti-thrombotic effects In macrophage-based inflammation assays magnolol inhibits NO production with reported IC50 values in the low micromolar range It serves as a suitable investigative compound for biomedical studies involving inflammation modulation infectious diseases and thrombosis research
Catalog No. 50-000-63274