Learn More
ATCC Magnusiomyces capitatus (de Hoog et al.) de Hoog et Smith - ATCC 10663 freeze-dried research strain
Supplier: ATCC 10663
Magnusiomyces capitatus is a freeze-dried fungal strain provided for laboratory research and reference use. The material contains viable cells that may include spores and mycelia and is intended for propagation and study by trained personnel.
- Freeze-dried format for improved stability during storage and transport.
- Store at 2°C to 8°C to preserve viability.
- Contains viable fungal cells, which may include spores and mycelia.
- Provided for laboratory research use only; not for therapeutic, diagnostic, or consumption use.
- Suitable for propagation, identification, and reference laboratory studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.