Learn More
Amsbio LLC MagSi-proteomics C18
Supplier:  Amsbio LLC MD01009
Encompass
MagSi-proteomics beads are magnetic silica beads coated with C4, C8 or C18 groups, providing a reversed phase (RP) surface chemistry. The beads are an ideal tool for protein and peptide sample concentration, desalting and fractionation, and reducing sample complexity.
Catalog No. 50-204-1474