Learn More
IBA LifeSciences MagStrep Strep-TactinXT Beads, 50 ml, magnetic beads, Strep-TactinXT coating, 5% (v/v) suspension, 20-40 μm size, 0.85 nmol/μl binding capacity
Supplier:  IBA LifeSciences 25090050
Encompass_Preferred
MagStrep Strep-TactinXT Beads, 50 ml, magnetic beads, Strep-TactinXT coating, 5% (v/v) suspension, 20-40 μm size, 0.85 nmol/μl binding capacity
Catalog No. NC3566339