Learn More
MSC Mahr Dial Indicator Back, Vertical Lug, Offset, Group 2
Supplier:  MSC 66002114
Encompass
This product is a Mahr dial indicator back designed with a vertical lug and offset configuration. It is primarily for ANSI/AGD Group 2 dial indicators but is also compatible with ANSI/AGD Group 1, 3, and 4 dial indicators, making it suitable for a range of measurement applications.
  • Vertical lug design for secure mounting.
  • Offset configuration for versatile positioning.
  • Compatible with ANSI/AGD Group 1, 2, 3, and 4 dial indicators.
  • Suitable for various measurement applications.
Catalog No. 50-005-27850