Learn More
Precision Plus Vacuum Parts Inc Major Repair Kit for Pfeiffer DUO 10C
Supplier:  Precision Plus Vacuum Parts Inc PKE32004T
Encompass_Preferred
Major Repair Kit includes o-rings, shaft seals, leak nozzle, hydraulic pistons, sight glass, sight glass gasket, springs, valves, and vanes for major pump maintenance
Catalog No. 50-195-6249