Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MAK683-CH2CH2COOH hydrochloride | 00-00-0 | 99.6% | 484.91 g·mol⁻¹ | C23H22ClFN6O3 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY130815A5MG

Encompass_Preferred

MAK683-CH2CH2COOH hydrochloride is a PROTAC linker supplied as a high-purity solid for research use. It binds to the EED protein and has been used with an E3 ligase ligand to design PROTAC degraders. Handle and store under recommended conditions to preserve stability and activity.

  • Binds to EED (embryonic ectoderm development protein).
  • High purity: 99.6%.
  • Molecular weight: 484.91 g·mol⁻¹.
  • Physical form: solid, white to light yellow.
  • Storage: keep refrigerated at 4°C and protect from light; store under inert gas for long-term stability.
  • Available in small milligram packages for research workflows.

Catalog No. 50-002-63007


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.