Learn More
Medchemexpress LLC Makisterone A | 20137-14-8 | 96.6% | 1 MG

Supplier: Medchemexpress LLC HY1280301MG
Makisterone A is a 28-carbon moulting hormone, identified as the major free pupal ecdysteroid in the honey bee, Apis mellifera.
- Molecular weight: 494.66
- Chemical formula: C28H46O7
- Appearance: Solid
- Color: White to off-white
- Soluble in DMSO
- Steroid structure
- Derived from Chenopodium quinoa Willd.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.