Learn More
Medchemexpress LLC Mal-amide-PEG8-Val-Ala-PAB-PNP | 2210247-59-7 | 95.7% | 1033.08 | C48H68N6O19 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY14842525MG
Encompass_Preferred
Mal-amide-PEG8-Val-Ala-PAB-PNP is a cleavable antibody-drug conjugate (ADC) linker used to attach cytotoxic payloads to antibodies and enable enzymatic release inside target cells. It combines a maleimide reactive group, a PEG8 spacer, a Val-Ala protease-cleavable dipeptide, a para-aminobenzyl self-immolative spacer, and a p-nitrophenyl carbonate leaving group.
  • Cleavable by proteases to enable intracellular payload release.
  • Maleimide group enables thiol conjugation to antibody cysteines.
  • PEG8 spacer improves solubility and linker flexibility.
  • High purity suitable for research bioconjugation (~95.7%).
  • Available in small mg-scale packs for laboratory conjugation workflows.
Catalog No. 50-002-02551