Learn More
Medchemexpress LLC Mal-amido-PEG12-NHS ester | 2101722-60-3 | 98.52% | C38H63N3O19 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY140977
Encompass_Preferred
Mal-amido-PEG12-NHS ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-003-52688