Learn More
Medchemexpress LLC Mal-amido-PEG2-C2-acid | 756525-98-1 | 99.8% | 328.32 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY42151
Encompass_Preferred
Mal-amido-PEG2-C2-acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs utilize the ubiquitin-proteasome system to selectively degrade target proteins
Catalog No. 50-004-02690