Learn More
Medchemexpress LLC Mal-amido-PEG2-C2-acid | 756525-98-1 | 99.8% | 328.32 | 5 G
SDP
Supplier:  Medchemexpress LLC HY421515G
Encompass_Preferred
Mal-amido-PEG2-C2-acid is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. PROTACs are compounds that utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting a ligand for an E3 ubiquitin ligase with a ligand for the target protein via a linker.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Enables selective degradation of target proteins
Catalog No. 50-004-01612