Learn More
TARGETMOL CHEMICALS INC MAL-AMIDO-PEG2-C2-AMIDO- 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1648610MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Mal-amido-PEG2-C2-amido-Ph-C2-CO-AZD (PF-05231023) is a long-acting fibroblast growth factor 21 (FGF21) analog and is an FGF21-receptor agonist. It is also suitable for development as a potential treatment for T2DM. purity: 99%
Catalog No. 50-257-7336