Learn More
Medchemexpress LLC Mal-amido-PEG2-C2-amido-Ph-C2-CO-AZD | 1037589-69-7 | 99.8% | 528.55 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY113697100MG
Encompass_Preferred
Mal-amido-PEG2-C2-amido-Ph-C2-CO-AZD is a polyethylene glycol (PEG)-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs are compounds designed to exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Contains two different ligands connected by a linker.
  • One ligand is for an E3 ubiquitin ligase.
  • The other ligand is for the target protein.
  • Utilizes the intracellular ubiquitin-proteasome system.
  • Selectively degrades target proteins.
Catalog No. 50-004-38016